Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 8 - 9 | 1535 | 89 | |
GenBank mRNA | AK032416.1 | - | 8 - 9 | 1498 | 89 | |
AK076799.1 | - | 8 - 9 | 1108 | 89 | ||
AK134544.1 | - | 5 - 6 | 402 | 89 | ||
AK141443.1 | - | 8 - 9 | 1085 | 89 | ||
AK150941.1 | - | 7 - 8 | 1010 | 89 | ||
AK151018.1 | - | 8 - 9 | 1505 | 89 | ||
AK152024.1 | - | 8 - 9 | 1505 | 89 | ||
AK152435.1 | - | 7 - 8 | 1010 | 89 | ||
AK172567.1 | - | 8 - 9 | 1501 | 89 | ||
AK173313.1 | - | 8 - 9 | 1154 | 89 | ||
BC010833.1 | - | 8 - 9 | 1150 | 4235165 | 89 | |
BC054121.1 | - | 8 - 9 | 1521 | 6393910 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |