Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026452.2 | NP_080728.1 | 7 - 8 | 861 | 95 | |
GenBank mRNA | AK004527.1 | - | 3 - 4 | 286 | 95 | |
AK009179.1 | - | 7 - 8 | 873 | 95 | ||
AK154343.1 | - | 7 - 8 | 886 | 95 | ||
AK158979.1 | - | 7 - 8 | 880 | 95 | ||
BC036386.1 | - | 7 - 8 | 861 | 1067021 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |