Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Katnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028805.2 | NP_083081.2 | 14 - 15 | 1665 | 72 | |
GenBank mRNA | AK010364.1 | - | 14 - 15 | 1596 | 72 | |
AK031630.1 | - | 14 - 15 | 1577 | 72 | ||
AK034103.1 | - | 14 - 15 | 1585 | 72 | ||
AK037858.1 | - | 1823 | 72 | |||
AK049345.1 | - | 14 - 15 | 1579 | 72 | ||
AK087973.1 | - | 14 - 15 | 1578 | 72 | ||
BC024500.1 | - | 5 - 6 | 472 | 72 | ||
BC045200.1 | - | 13 - 14 | 1576 | 5322489 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |