Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028500.3 | NP_082776.2 | 3 - 4 | 492 | 139 | |
GenBank mRNA | AB206786.1 | - | 3 - 4 | 397 | 139 | |
AK006582.1 | - | 3 - 4 | 493 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |