Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fn3krp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181420.3 | NP_852085.2 | 1 - 2 | 236 | 89 | |
GenBank mRNA | AY360466.1 | - | 1 - 2 | 194 | 89 | |
BC032265.1 | - | 1 - 2 | 182 | 4015688 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |