Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177699.4 | NP_808367.2 | 5 - 6 | 665 | 62 | |
GenBank mRNA | AB041045.1 | - | 5 - 6 | 559 | 62 | |
AK030737.1 | - | 5 - 6 | 627 | 62 | ||
BC017144.1 | - | 4 - 5 | 676 | 62 | ||
BC060654.1 | - | 5 - 6 | 648 | 30354865 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |