Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10579654 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10579651 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10579655 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10579650 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10579653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10579652 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1444438_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10579649 |
- |
- |
Mouse Genome 430 2.0 Array |
1444438_at |
- |
- |
Mouse Genome 430 2.0 Array |
1444438_PM_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
234421_at |
- |
- |
Affymetrix GeneChip Mouse Genome 430 2.0 Array |
234421_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800018766.mm.1 |
NM_001080812.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018766.mm.1 |
NM_001080812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018766.mm.1 |
TC0800002452.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800018767.mm.1 |
NM_001080812.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018767.mm.1 |
NM_001080812.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800018767.mm.1 |
TC0800002452.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1444438_PM_at |
- |
- |
Mouse Expression Set 430 |
1444438_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444438_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1444438_PM_at |
- |
- |