Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001146059.1 | NP_001139531.1 | 7 - 8 | 861 | 111 | |
NM_001146060.1 | NP_001139532.1 | 7 - 8 | 941 | 111 | ||
NM_146228.4 | NP_666340.3 | 7 - 8 | 858 | 111 | ||
GenBank mRNA | AB107016.1 | - | 7 - 8 | 858 | 111 | |
AF481142.1 | - | 6 - 7 | 754 | 111 | ||
AK053076.1 | - | 7 - 8 | 831 | 111 | ||
AK131149.1 | - | 7 - 8 | 860 | 111 | ||
AK155451.1 | - | 7 - 8 | 927 | 111 | ||
BC117928.1 | - | 6 - 7 | 811 | 40097635 | 111 | |
BC117929.1 | - | 6 - 7 | 808 | 40097636 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |