Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083319.1 | NP_001076788.1 | 14 - 15 | 1799 | 75 | |
NM_013699.2 | NP_038727.1 | 13 - 14 | 1691 | 75 | ||
GenBank mRNA | AB221581.1 | - | 12 - 13 | 1279 | 75 | |
AK079815.1 | - | 6 - 7 | 526 | 75 | ||
AK140942.1 | - | 8 - 9 | 696 | 75 | ||
AK146040.1 | - | 2 - 3 | 311 | 75 | ||
AY182062.1 | - | 13 - 14 | 1388 | 75 | ||
BC034749.1 | - | 12 - 13 | 2018 | 75 | ||
BC139828.1 | - | 12 - 13 | 1308 | 40142981 | 75 | |
BC141082.1 | - | 12 - 13 | 1370 | 9055694 | 75 | |
U20086.1 | - | 12 - 13 | 1282 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |