Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198862.2 | NP_942562.2 | 2 - 3 | 916 | 59 | |
GenBank mRNA | AK136883.1 | - | 3 - 4 | 1482 | 59 | |
AK173159.1 | - | 2 - 3 | 916 | 59 | ||
BE952199.1 | - | 2 - 3 | 120 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |