Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026482.2 | NP_080758.1 | 10 - 11 | 2032 | 125 | |
GenBank mRNA | BC029045.1 | - | 4 - 5 | 672 | 125 | |
BC030901.1 | - | 4 - 5 | 673 | 125 | ||
BC049262.1 | - | 4 - 5 | 672 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |