Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011403.2 | NP_035533.1 | 1 - 2 | 225 | 68 | |
GenBank mRNA | AK137542.1 | - | 1 - 2 | 121 | 68 | |
AK157181.1 | - | 1 - 2 | 194 | 68 | ||
AK157833.1 | - | 1 - 2 | 83 | 68 | ||
AK157861.1 | - | 1 - 2 | 95 | 68 | ||
AK158143.1 | - | 1 - 2 | 226 | 68 | ||
AK158184.1 | - | 1 - 2 | 191 | 68 | ||
AK158243.1 | - | 1 - 2 | 95 | 68 | ||
AK158453.1 | - | 1 - 2 | 70 | 68 | ||
AK162616.1 | - | 1 - 2 | 68 | |||
AK165195.1 | - | 1 - 2 | 82 | 68 | ||
AK165267.1 | - | 1 - 2 | 82 | 68 | ||
AK171662.1 | - | 121 | 68 | |||
AK171808.1 | - | 1 - 2 | 59 | 68 | ||
AK171845.1 | - | 121 | 68 | |||
AK172103.1 | - | 1 - 2 | 84 | 68 | ||
AK172316.1 | - | 1 - 2 | 70 | 68 | ||
AK172395.1 | - | 1 - 2 | 121 | 68 | ||
AK172456.1 | - | 1 - 2 | 107 | 68 | ||
AK172569.1 | - | 1 - 2 | 70 | 68 | ||
AY296129.1 | - | 1 - 2 | 58 | 68 | ||
BC052419.1 | - | 1 - 2 | 65 | 5702599 | 68 | |
BC053429.1 | - | 1 - 2 | 56 | 6506270 | 68 | |
M29379.1 | - | 1 - 2 | 52 | 68 | ||
X02677.1 | - | 1 - 2 | 58 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |