Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200041680.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041680.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041680.mm.1 |
TC0200004913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041686.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041686.mm.1 |
NM_001289717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041686.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041686.mm.1 |
TC0200004913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041675.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041675.mm.1 |
NM_001289717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041675.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041675.mm.1 |
NM_009743.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041675.mm.1 |
TC0200004913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041681.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041681.mm.1 |
NM_001289717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041681.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041681.mm.1 |
TC0200004913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041674.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041674.mm.1 |
NM_001289717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041674.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041674.mm.1 |
NM_009743.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041674.mm.1 |
TC0200004913.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200041676.mm.1 |
NM_001289716.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041676.mm.1 |
NM_001289717.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041676.mm.1 |
NM_001289739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041676.mm.1 |
NM_009743.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200041676.mm.1 |
TC0200004913.mm.1 |
MTA TC |