Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161329.1 | NP_001154801.1 | 15 - 16 | 1919 | 87 | |
NM_133702.2 | NP_598463.2 | 15 - 16 | 1979 | 87 | ||
GenBank mRNA | AK038696.1 | - | 15 - 16 | 1810 | 87 | |
AK076704.1 | - | 15 - 16 | 1920 | 87 | ||
AK078524.1 | - | 15 - 16 | 1870 | 87 | ||
AK157743.1 | - | 15 - 16 | 1919 | 87 | ||
BC004785.1 | - | 10 - 11 | 1258 | 87 | ||
BC024874.1 | - | 15 - 16 | 1858 | 3984027 | 87 | |
BC079576.1 | - | 1 - 2 | 1013 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |