Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcakd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026551.3 | NP_080827.2 | 3 - 4 | 508 | 100 | |
GenBank mRNA | AK051022.1 | - | 3 - 4 | 509 | 100 | |
AK078460.1 | - | 3 - 4 | 441 | 100 | ||
AK162069.1 | - | 3 - 4 | 468 | 100 | ||
AK170762.1 | - | 3 - 4 | 459 | 100 | ||
BC005607.1 | - | 3 - 4 | 474 | 3498151 | 100 | |
BC023208.1 | - | 2 - 3 | 422 | 100 | ||
BC031855.1 | - | 3 - 4 | 457 | 100 | ||
BC042685.1 | - | 2 - 3 | 422 | 100 | ||
BC045193.1 | - | 3 - 4 | 462 | 5368157 | 100 | |
BC047340.1 | - | 2 - 3 | 422 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |