Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112705.2 | NP_001106176.1 | 18 - 19 | 2247 | 126 | |
NM_001294329.1 | NP_001281258.1 | 16 - 17 | 2107 | 126 | ||
NM_001294330.1 | NP_001281259.1 | 16 - 17 | 2055 | 126 | ||
NM_001294331.1 | NP_001281260.1 | 18 - 19 | 2260 | 126 | ||
NM_001294334.1 | NP_001281263.1 | 18 - 19 | 2247 | 126 | ||
GenBank mRNA | AF045252.1 | - | 18 - 19 | 2266 | 126 | |
AF045253.1 | - | 17 - 18 | 1694 | 126 | ||
AF045254.1 | - | 13 - 14 | 1408 | 126 | ||
AK089925.1 | - | 16 - 17 | 1635 | 126 | ||
AK165667.1 | - | 18 - 19 | 2250 | 126 | ||
AK211763.1 | - | 2 - 3 | 241 | 126 | ||
AX775743.1 | - | 12 - 13 | 1272 | 126 | ||
BC066198.1 | - | 16 - 17 | 1600 | 126 | ||
U56909.1 | - | 9 - 10 | 945 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |