Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021546.3 | NP_067521.2 | 7 - 8 | 627 | 96 | |
GenBank mRNA | AB039948.1 | - | 5 - 6 | 448 | 96 | |
AK013520.1 | - | 7 - 8 | 626 | 96 | ||
AK044296.1 | - | 7 - 8 | 619 | 96 | ||
AK158225.1 | - | 7 - 8 | 629 | 96 | ||
BC126874.1 | - | 5 - 6 | 441 | 40044848 | 96 | |
BC126875.1 | - | 5 - 6 | 441 | 40044849 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |