Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291111.1 | NP_001278040.1 | 7 - 8 | 758 | 96 | |
NM_008180.2 | NP_032206.1 | 8 - 9 | 893 | 96 | ||
GenBank mRNA | AK033435.1 | - | 2 - 3 | 1368 | 96 | |
AK146486.1 | - | 8 - 9 | 798 | 96 | ||
AK149577.1 | - | 7 - 8 | 663 | 96 | ||
AK159630.1 | - | 8 - 9 | 818 | 96 | ||
AK169096.1 | - | 8 - 9 | 813 | 96 | ||
BC003784.1 | - | 8 - 9 | 806 | 3593913 | 96 | |
U35456.1 | - | 7 - 8 | 789 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |