Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_001139511.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_001139511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_001139513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_001139513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_023130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
NM_023130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018696.mm.1 |
TC0200002352.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_001139511.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_001139511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_001139513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_001139513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_023130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
NM_023130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018701.mm.1 |
TC0200002352.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_001139511.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_001139511.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_001139513.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_001139513.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_023130.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
NM_023130.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200018702.mm.1 |
TC0200002352.mm.1 |
MTA TC |