Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019642.4 | NP_062616.2 | 1 - 2 | 58 | ||
GenBank mRNA | AK004968.1 | - | 1 - 2 | 58 | ||
AK018761.2 | - | 1 - 2 | 58 | |||
AK161624.1 | - | 1 - 2 | 58 | |||
BC046806.1 | - | 1 - 2 | 6829209 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |