Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200042473.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042473.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042473.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042473.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042473.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042468.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042468.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042468.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042468.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042468.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042475.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042475.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042475.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042475.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042475.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042466.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042466.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042466.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042466.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042466.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042467.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042467.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042467.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042467.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042467.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042464.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042464.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042464.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042464.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042464.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200042476.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042476.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042476.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042476.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200042476.mm.1 |
TC0200005511.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200046701.mm.1 |
NM_170588.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046701.mm.1 |
NM_170588.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046701.mm.1 |
NM_170590.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046701.mm.1 |
NM_170590.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200046701.mm.1 |
TC0200005511.mm.1 |
MTA TC |