Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sla2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029983.5 | NP_084259.1 | 6 - 7 | 757 | 90 | |
GenBank mRNA | AF287467.1 | - | 6 - 7 | 808 | 90 | |
AF434990.1 | - | 6 - 7 | 800 | 90 | ||
AK020837.1 | - | 4 - 5 | 414 | 90 | ||
AK030877.1 | - | 6 - 7 | 758 | 90 | ||
AK088672.1 | - | 6 - 7 | 884 | 90 | ||
AK138709.1 | - | 6 - 7 | 758 | 90 | ||
BC052655.1 | - | 6 - 7 | 856 | 30040401 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |