Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291190.1 | NP_001278119.1 | 7 - 8 | 893 | 127 | |
NM_177710.4 | NP_808378.2 | 8 - 9 | 757 | 127 | ||
GenBank mRNA | AB099288.1 | - | 8 - 9 | 757 | 127 | |
AK155232.1 | - | 7 - 8 | 884 | 127 | ||
AK170142.1 | - | 7 - 8 | 890 | 127 | ||
BC141392.1 | - | 8 - 9 | 751 | 9056005 | 127 | |
BC141393.1 | - | 8 - 9 | 751 | 9056006 | 127 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons |