Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100035605.mm.1 |
NM_001098836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035605.mm.1 |
NM_001098836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035605.mm.1 |
NM_001098837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035605.mm.1 |
NM_001098837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035605.mm.1 |
TC1100003830.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035602.mm.1 |
NM_001098836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035602.mm.1 |
NM_001098836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035602.mm.1 |
NM_001098837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035602.mm.1 |
NM_001098837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035602.mm.1 |
TC1100003830.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035601.mm.1 |
NM_001098836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035601.mm.1 |
NM_001098836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035601.mm.1 |
NM_001098837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035601.mm.1 |
NM_001098837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035601.mm.1 |
TC1100003830.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100035603.mm.1 |
NM_001098836.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035603.mm.1 |
NM_001098836.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035603.mm.1 |
NM_001098837.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035603.mm.1 |
NM_001098837.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100035603.mm.1 |
TC1100003830.mm.1 |
MTA TC |