Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atxn7l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098836.1 | NP_001092306.1 | 3 - 4 | 471 | 93 | |
NM_001098837.1 | NP_001092307.1 | 3 - 4 | 471 | 93 | ||
GenBank mRNA | AK090368.1 | - | 3 - 4 | 473 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |