Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009053.2 | NP_033079.1 | 2 - 3 | 367 | 59 | |
GenBank mRNA | AF015770.1 | - | 2 - 3 | 325 | 59 | |
AK004573.1 | - | 2 - 3 | 350 | 59 | ||
AK207192.1 | - | 1 - 2 | 54 | 59 | ||
AK218494.1 | - | 1 - 2 | 35 | 59 | ||
BC066023.1 | - | 2 - 3 | 367 | 6829176 | 59 | |
U94350.1 | - | 2 - 3 | 360 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |