Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038653.1 | NP_001033742.1 | 4 - 5 | 564 | 79 | |
NM_001038654.1 | NP_001033743.1 | 3 - 4 | 469 | 79 | ||
NM_030696.3 | NP_109621.1 | 4 - 5 | 576 | 79 | ||
GenBank mRNA | AF178954.1 | - | 2 - 3 | 364 | 79 | |
AK149707.1 | - | 4 - 5 | 567 | 79 | ||
AK149983.1 | - | 4 - 5 | 567 | 79 | ||
AK165596.1 | - | 3 - 4 | 412 | 79 | ||
AK166048.1 | - | 552 | 79 | |||
AK169917.1 | - | 4 - 5 | 526 | 79 | ||
AK171260.1 | - | 4 - 5 | 552 | 79 | ||
BC046525.1 | - | 4 - 5 | 576 | 5319754 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |