Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008682.2 | NP_032708.2 | 6 - 7 | 828 | 115 | |
GenBank mRNA | AK010591.1 | - | 6 - 7 | 816 | 115 | |
AK048393.1 | - | 6 - 7 | 780 | 115 | ||
AK085355.1 | - | 6 - 7 | 791 | 115 | ||
AK090040.1 | - | 6 - 7 | 773 | 115 | ||
AK153746.1 | - | 6 - 7 | 830 | 115 | ||
BC052430.1 | - | 6 - 7 | 797 | 6836583 | 115 | |
BC066870.1 | - | 6 - 7 | 765 | 30476383 | 115 | |
D10712.1 | - | 6 - 7 | 770 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |