Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011936.2 | NP_036066.2 | 2 - 3 | 176 | 103 | |
GenBank mRNA | AJ237917.1 | - | 2 - 3 | 162 | 103 | |
AK036677.1 | - | 2 - 3 | 166 | 103 | ||
AK040866.1 | - | 2 - 3 | 154 | 103 | ||
AK045465.1 | - | 2 - 3 | 153 | 103 | ||
AK046935.1 | - | 2 - 3 | 135 | 103 | ||
AK049502.1 | - | 2 - 3 | 169 | 103 | ||
AK088881.1 | - | 2 - 3 | 162 | 103 | ||
AK129437.1 | - | 2 - 3 | 143 | 103 | ||
AK161060.1 | - | 2 - 3 | 135 | 103 | ||
BC022222.1 | - | 2 - 3 | 127 | 4237261 | 103 | |
BC057008.1 | - | 2 - 3 | 142 | 5708558 | 103 | |
BY290198.1 | - | 2 - 3 | 177 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |