Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198839.1 | NP_001185768.1 | 1 - 2 | 97 | 72 | |
GenBank mRNA | AK138372.1 | - | 1 - 2 | 67 | 72 | |
AK159168.1 | - | 1 - 2 | 64 | 72 | ||
AK159297.1 | - | 1 - 2 | 64 | 72 | ||
AK167352.1 | - | 1 - 2 | 98 | 72 | ||
AK167504.1 | - | 1 - 2 | 98 | 72 | ||
U28151.1 | - | 1 - 2 | 71 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |