Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130868.2 | NP_001124340.1 | 3 - 4 | 479 | 59 | |
NM_001286384.1 | NP_001273313.1 | 4 - 5 | 537 | 59 | ||
NM_053092.3 | NP_444322.1 | 2 - 3 | 299 | 59 | ||
GenBank mRNA | AK089053.1 | - | 2 - 3 | 224 | 59 | |
AK090203.1 | - | 2 - 3 | 224 | 59 | ||
AK153236.1 | - | 2 - 3 | 222 | 59 | ||
AK167693.1 | - | 2 - 3 | 224 | 59 | ||
BC027356.1 | - | 4 - 5 | 449 | 4950333 | 59 | |
BC035324.1 | - | 3 - 4 | 391 | 2811286 | 59 | |
BC036289.1 | - | 2 - 3 | 268 | 3157706 | 59 | |
BY792549.1 | - | 2 - 3 | 250 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |