Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000019372.mm.1 |
NM_029102.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019372.mm.1 |
NM_029102.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019372.mm.1 |
TC1000002571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019371.mm.1 |
NM_029102.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019371.mm.1 |
NM_029102.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019371.mm.1 |
TC1000002571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019366.mm.1 |
NM_029102.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019366.mm.1 |
NM_029102.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019366.mm.1 |
TC1000002571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019368.mm.1 |
NM_029102.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019368.mm.1 |
NM_029102.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019368.mm.1 |
TC1000002571.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000019373.mm.1 |
NM_029102.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019373.mm.1 |
NM_029102.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000019373.mm.1 |
TC1000002571.mm.1 |
MTA TC |