Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172760.3 | NP_766348.3 | 3 - 4 | 349 | 64 | |
GenBank mRNA | AK037067.1 | - | 3 - 4 | 342 | 64 | |
BC058752.1 | - | 3 - 4 | 332 | 6485393 | 64 | |
BC138887.1 | - | 3 - 4 | 306 | 8861909 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |