Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033320.2 | NP_001028492.1 | 12 - 13 | 1964 | 62 | |
GenBank mRNA | AK031012.1 | - | 12 - 13 | 1963 | 62 | |
HF678090.1 | - | 33 - 34 | 3595 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |