Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033320.2 | NP_001028492.1 | 1 - 2 | 339 | 69 | |
GenBank mRNA | AK031012.1 | - | 1 - 2 | 338 | 69 | |
HF678090.1 | - | 22 - 23 | 1970 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |