Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rltpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033320.2 | NP_001028492.1 | 2 - 3 | 443 | 76 | |
GenBank mRNA | AK031012.1 | - | 2 - 3 | 442 | 76 | |
HF678090.1 | - | 23 - 24 | 2074 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |