Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmtc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177368.4 | NP_796342.2 | 9 - 10 | 2883 | 79 | |
GenBank mRNA | BC034657.1 | - | 2 - 3 | 104 | 79 | |
BC090655.1 | - | 9 - 10 | 2245 | 79 | ||
BC092226.1 | - | 9 - 10 | 2839 | 6852756 | 79 | |
BC138362.1 | - | 9 - 10 | 2614 | 8861382 | 79 | |
BC138363.1 | - | 9 - 10 | 2614 | 8861383 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |