Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025575.3 | NP_079851.2 | 2 - 3 | 392 | 82 | |
GenBank mRNA | AK003071.1 | - | 2 - 3 | 385 | 82 | |
AK005249.1 | - | 2 - 3 | 295 | 82 | ||
AK005510.1 | - | 2 - 3 | 329 | 82 | ||
AK007882.1 | - | 2 - 3 | 391 | 82 | ||
AK009684.1 | - | 2 - 3 | 356 | 82 | ||
AK011753.1 | - | 2 - 3 | 388 | 82 | ||
AK013220.1 | - | 2 - 3 | 365 | 82 | ||
AK032073.1 | - | 2 - 3 | 363 | 82 | ||
AK032098.1 | - | 1 - 2 | 227 | 82 | ||
AK075770.1 | - | 2 - 3 | 357 | 82 | ||
AK148305.1 | - | 2 - 3 | 330 | 82 | ||
BC038166.1 | - | 2 - 3 | 82 | |||
BC053943.1 | - | 1 - 2 | 226 | 82 | ||
BC054055.1 | - | 2 - 3 | 82 | |||
BC061084.1 | - | 1 - 2 | 266 | 6596164 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |