Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfdn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013369.2 | NP_001013387.1 | 2 - 3 | 279 | 71 | |
NM_001110152.2 | NP_001103622.1 | 2 - 3 | 347 | 71 | ||
GenBank mRNA | AK007940.1 | - | 2 - 3 | 141 | 71 | |
AK136249.1 | - | 2 - 3 | 348 | 71 | ||
BC065407.1 | - | 2 - 3 | 279 | 71 | ||
BU503885.1 | - | 1 - 2 | 118 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |