Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025988.2 | NP_080264.1 | 5 - 6 | 848 | 109 | |
GenBank mRNA | AK008170.1 | - | 4 - 5 | 381 | 109 | |
AK008243.1 | - | 3 - 4 | 397 | 109 | ||
AK079604.1 | - | 4 - 5 | 717 | 109 | ||
AK148438.1 | - | 343 | 109 | |||
BC048371.1 | - | 4 - 5 | 762 | 6396076 | 109 | |
BI734653.1 | - | 4 - 5 | 382 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |