Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pitpnm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024927.2 | NP_001020098.1 | 17 - 18 | 2523 | 70 | |
NM_001081641.1 | NP_001075110.1 | 16 - 17 | 2475 | 70 | ||
GenBank mRNA | AK147444.1 | - | 17 - 18 | 2480 | 70 | |
AK147630.1 | - | 16 - 17 | 2308 | 70 | ||
BC112411.1 | - | 13 - 14 | 2437 | 40061037 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |