Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK081566.1 | - | 12 - 13 | 1378 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |