Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026824.4 | NP_081100.2 | 7 - 8 | 955 | 84 | |
GenBank mRNA | AK004041.1 | - | 6 - 7 | 478 | 84 | |
AK037538.1 | - | 5 - 6 | 2092 | 84 | ||
AK088243.1 | - | 7 - 8 | 956 | 84 | ||
AK147967.1 | - | 7 - 8 | 1154 | 84 | ||
AK148307.1 | - | 7 - 8 | 1154 | 84 | ||
BC019480.1 | - | 5 - 6 | 439 | 4037583 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |