Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sectm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145373.2 | NP_663348.2 | 1 - 2 | 54 | 64 | |
GenBank mRNA | AK082020.1 | - | 1 - 2 | 55 | 64 | |
AK170159.1 | - | 1 - 2 | 50 | 64 | ||
AK171246.1 | - | 1 - 2 | 56 | 64 | ||
AW985540.1 | - | 1 - 2 | 54 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |