Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113486.1 | NP_001106958.1 | 2 - 3 | 172 | 113 | |
GenBank mRNA | AK141312.1 | - | 2 - 3 | 113 | 113 | |
BC046524.1 | - | 2 - 3 | 172 | 5290621 | 113 | |
BC094455.1 | - | 2 - 3 | 95 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |