Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026364.1 | NP_080640.1 | 4 - 5 | 589 | 66 | |
GenBank mRNA | AK011290.1 | - | 4 - 5 | 559 | 66 | |
AK077439.1 | - | 4 - 5 | 592 | 66 | ||
AK177728.1 | - | 2 - 3 | 223 | 66 | ||
AK183483.1 | - | 1 - 2 | 116 | 66 | ||
AK208020.1 | - | 1 - 2 | 116 | 66 | ||
AK218181.1 | - | 4 - 5 | 221 | 66 | ||
BC029621.1 | - | 4 - 5 | 580 | 4208743 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |