Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177449.3 | NP_803232.2 | 1 - 2 | 45 | 73 | |
GenBank mRNA | AK148120.1 | - | 1 - 2 | 47 | 73 | |
AK170400.1 | - | 1 - 2 | 45 | 73 | ||
CB236245.1 | - | 1 - 2 | 48 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |