Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053273.2 | NP_444503.2 | 9 - 10 | 1101 | 56 | |
GenBank mRNA | AK152172.1 | - | 9 - 10 | 1053 | 56 | |
AK154835.1 | - | 9 - 10 | 1108 | 56 | ||
AK155166.1 | - | 9 - 10 | 1102 | 56 | ||
AK156393.1 | - | 9 - 10 | 1054 | 56 | ||
AK168408.1 | - | 9 - 10 | 1054 | 56 | ||
AK204126.1 | - | 2 - 3 | 143 | 56 | ||
BC060740.1 | - | 9 - 10 | 1044 | 4158813 | 56 | |
BY731753.1 | - | 5 - 6 | 412 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |