Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC021835.1 | - | 7 - 8 | 1701 | 83 | |
EU887626.1 | - | 10 - 11 | 3185 | 83 | ||
EU887629.1 | - | 10 - 11 | 3209 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |