Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037809.5 | NP_001032898.1 | 14 - 15 | 2215 | 87 | |
NM_007665.3 | NP_031691.1 | 14 - 15 | 2212 | 87 | ||
GenBank mRNA | AK031265.1 | - | 14 - 15 | 2155 | 87 | |
AK045041.1 | - | 14 - 15 | 2175 | 87 | ||
AK164577.1 | - | 14 - 15 | 2185 | 87 | ||
BC052189.1 | - | 12 - 13 | 1903 | 87 | ||
BC098459.1 | - | 14 - 15 | 2121 | 30079668 | 87 | |
X06340.1 | - | 14 - 15 | 2153 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |